missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyethyl Methacrylate, TRC

Product Code. 30138398
Change view
Click to view available options
Quantity:
500 mg
1 g
25 g
Unit Size:
1g
25g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138398 25 g 25g
30135305 1 g 1g
30010022 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138398 Supplier Toronto Research Chemicals Supplier No. TRCH94223025G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868-77-9
Molecular Formula C6 H10 O3
Molecular Weight (g/mol) 130.14
Synonym 2-Propenoic acid, 2-methyl-, 2-hydroxyethyl ester, Methacrylic acid, 2-hydroxyethyl ester (6CI,8CI), Methacrylic acid, ester with glycol (7CI), 2-(Methacryloyloxy)ethanol, 2-HEMA, 2-Hydroxyethyl 2-methylprop-2-enoate, 2-Hydroxyethyl methacrylate, Acryester HISS, Acryester HO, Acryester HOMA, Acryl Ester HO, Bisomer HEMA, Bisomer SR, Blemmer E, EB 109, EB 109 (monomer), EM 321, Ethylene glycol methacrylate, Ethylene glycol monomethacrylate, GE 610, Glycol methacrylate, Glycol monomethacrylate, HEMA, HEMA 90, JB 4 Plus, Kayarad 2-HEMA, Light Ester HO, Light Ester HO 250, Light Ester HO 250M, Light Ester HO 250N, M 0085, Mhoromer BM 903, Mhoromer BM 905, Monomer MG 1, NSC 24180, Rocryl 400, Visiomer HEMA 98, β-Hydroxyethyl methacrylate
IUPAC Name 2-hydroxyethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCO

Specifications

Quantity 25 g
InChI Formula InChI=1S/C6H10O3/c1-5(2)6(8)9-4-3-7/h7H,1,3-4H2,2H3
Formula Weight 130.063
Chemical Name or Material 2-Hydroxyethyl Methacrylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.