missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyethyl methacrylate, 97%, stabilized

Product Code. 10071750 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
1kg
2.5kg
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10071750 500 g Glass bottle 500g
10488383 250 g Glass bottle 250g
10367900 1 kg Glass bottle 1kg
10559433 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10071750 Supplier Thermo Scientific Acros Supplier No. 156335000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 868-77-9 | C6H10O3 | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868-77-9 , 150-76-5
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00002863
InChI Key WOBHKFSMXKNTIM-UHFFFAOYSA-N
Synonym 2-hydroxyethyl methacrylate, glycol methacrylate, hydroxyethyl methacrylate, glycol monomethacrylate, hema, ethylene glycol methacrylate, 2-methacryloyloxy ethanol, 2-hydroxyethylmethacrylate, mhoromer, monomer mg-1
PubChem CID 13360
ChEBI CHEBI:34288
IUPAC Name 2-hydroxyethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCO

Specifications

Acidity 2mg KOH/g max.
CAS Min % 96.0
CAS Max % 100.0
Melting Point -12.0°C
Density 1.0730g/mL
Boiling Point 213°C
Flash Point 106°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CO2CH2CH2OH
Refractive Index 1.4510 to 1.4530
Quantity 500 g
Beilstein 02,III,1291
Specific Gravity 1.073
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most common organic solvents
Stabilizer 200 to 400ppm MEHQ
Viscosity 5 mPa.s (30°C)
Formula Weight 130.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Hydroxyethyl methacrylate
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxyethyl methacrylate, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Hydroxyethyl methacrylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.