missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyethyl methacrylate, 97%, stab. with 4-methoxyphenol

Product Code. p-7027477
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11481307

Brand: Thermo Scientific Alfa Aesar B24260.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 868-77-9 | C6H10O3 | 130.143 g/mol

2-Hydroxyethyl methacrylate is used in UV-curable inks and coatings. It is also used in adhesives, artificial nails, dental materials and lacquers. In dentistry, it is one of the main volatile acrylates along with methyl methacrylate. Further, it is used as a monomer in the synthesis of polymers for dental prosthetics and for geotechnical grouting in construction work.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxyethyl methacrylate is used in UV-curable inks and coatings. It is also used in adhesives, artificial nails, dental materials and lacquers. In dentistry, it is one of the main volatile acrylates along with methyl methacrylate. Further, it is used as a monomer in the synthesis of polymers for dental prosthetics and for geotechnical grouting in construction work.

Solubility
Slightly miscible with hot water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868-77-9
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00002863
InChI Key WOBHKFSMXKNTIM-UHFFFAOYSA-N
Synonym 2-hydroxyethyl methacrylate, glycol methacrylate, hydroxyethyl methacrylate, glycol monomethacrylate, hema, ethylene glycol methacrylate, 2-methacryloyloxy ethanol, 2-hydroxyethylmethacrylate, mhoromer, monomer mg-1
PubChem CID 13360
ChEBI CHEBI:34288
IUPAC Name 2-hydroxyethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCO

Specifications

Melting Point -12°C
Density 1.071
Boiling Point 250°C
Flash Point 101°C (213°F)
Odor Ester-like
Refractive Index 1.453
Quantity 100 g
Beilstein 1071583
Sensitivity Light sensitive
Solubility Information Slightly miscible with hot water.
Formula Weight 130.14
Percent Purity 97%
Chemical Name or Material 2-Hydroxyethyl methacrylate, stabilized with ≈500ppm 4-methoxyphenol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxyethyl methacrylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2-Hydroxyethyl methacrylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.