missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Hydroxyethyl)hydrazine, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
(2-Hydroxyethyl)hydrazine is used as a plant growth regulant, a component in jet fuels, intermediate for organic synthesis especially heterocycles used in agrochemicals, pharmaceuticals, stabilizers and polymerizations, Chain extender for urethane formulations, corrosion inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -70°C |
Density | 1.121 g/mL |
Boiling Point | 218°C to 220°C |
Flash Point | 73°C (163°F) |
Refractive Index | 1.493 |
Quantity | 250 g |
UN Number | UN2810 |
Solubility Information | Soluble in water,alcohols and glycols. |
Formula Weight | 76.1 |
Physical Form | Liquid |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'