missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyethanesulfonic Acid (80% w/w in Water), TRC

Product Code. 30408021
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30408021 50 g
30407328 10 g
30391661 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30408021 Supplier Toronto Research Chemicals Supplier No. TRCH55339550G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-36-8
Molecular Formula C2H5O4S
Molecular Weight (g/mol) 126.13
Synonym (2-Hydroxyethyl)sulfonic Acid, (Hydroxyethyl)sulfonic Acid, 2-Hydroxyethane-1-sulfonic Acid, Ethanolsulfonic Acid, Isethionic Acid, NSC 60516, Tego HES 70
IUPAC Name 2-hydroxyethanesulfonic acid
SMILES OCCS(=O)(=O)O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C2H6O4S/c3-1-2-7(4,5)6/h3H,1-2H2,(H,4,5,6)
Formula Weight 125.9987
Concentration or Composition (by Analyte or Components) 0.8
Chemical Name or Material 2-Hydroxyethanesulfonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.