missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyestriol, TRC

Product Code. 30396109
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396109 5 mg -
30399443 1 mg 1mg
30400065 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396109 Supplier Toronto Research Chemicals Supplier No. TRCH9493135MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1232-80-0
Molecular Formula C18H24O4
Molecular Weight (g/mol) 304.38
Synonym (16α,17β)-Estra-1,3,5(10)-triene-2,3,16,17-tetrol, 2-Hydroxy-16α,17β-estriol
IUPAC Name (8R,9S,13S,14S,16R,17R)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-2,3,16,17-tetrol
SMILES OC(C(O)=C1)=CC2=C1[C@@]3([H])CC[C@]4(C)[C@@H](O)[C@H](O)C[C@@]4([H])[C@]3([H])CC2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C18H24O4/c1-18-5-4-10-11(13(18)8-16(21)17(18)22)3-2-9-6-14(19)15(20)7-12(9)10/h6-7,10-11,13,16-17,19-22H,2-5,8H2,1H3/t10-,11+,13-,16+,17-,18-/m0/s1
Formula Weight 304.1675
Chemical Name or Material 2-Hydroxyestriol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.