missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxydiphenylmethane, 98%, Thermo Scientific™

Product Code. 10652002 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10652002

Brand: Thermo Scientific 162000050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28994-41-4
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.238
MDL Number MFCD00002242
InChI Key CDMGNVWZXRKJNS-UHFFFAOYSA-N
Synonym 2-hydroxydiphenylmethane, o-benzylphenol, phenol, 2-phenylmethyl, benzylphenol, 2-benzyl-phenol, 2-phenylmethyl phenol, alpha-phenyl-o-cresol, o-cresol, .alpha.-phenyl, unii-sf555bl24o, 2-benzyl phenol
PubChem CID 24216
IUPAC Name 2-benzylphenol
SMILES C1=CC=C(C=C1)CC2=CC=CC=C2O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 51°C to 55°C
Color Colorless to Pink-Red
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula C6H5CH2C6H4OH
Quantity 5 g
Formula Weight 184.24
Percent Purity ≥97.5%
Physical Form Crystals
Chemical Name or Material 2-Hydroxydiphenylmethane, 98%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.