missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxydecanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Hydroxydecanoic Acid |
| CAS | 5393-81-7 |
| Molecular Formula | C10H20O3 |
| Quantity | 100 mg |
| Synonym | (±)-2-Hydroxydecanoic Acid, (±)-alpha-Hydroxycapric Acid, (±)-alpha-Hydroxydecanoic Acid, 2-Hydroxycapric Acid, DL-2-Hydroxycapric Acid, NSC 4801, alpha-Hydroxycapric Acid, alpha-Hydroxydecanoic Acid |
| InChI Formula | InChI=1S/C10H20O3/c1-2-3-4-5-6-7-8-9(11)10(12)13/h9,11H,2-8H2,1H3,(H,12,13) |
| SMILES | CCCCCCCCC(O)C(=O)O |
| IUPAC Name | 2-hydroxydecanoic acid |
| Molecular Weight (g/mol) | 188.26 |
| Formula Weight | 188.1412 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?