missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxycyclohexanone dimer, 90+%

Product Code. 11392169
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392169 5 g 5g
11382169 1 g 1g
11302179 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11392169 Supplier Thermo Scientific Alfa Aesar Supplier No. H27199.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 30282-14-5 | C6H10O2 | 114.144 g/mol

It is a reactant involved in the synthesis of substrates for tin tetrachloride mediated fragmentation to produce ynones and ynoates, tandem oxidation coupling reactions with diamines, asymmetric allylic alkylation of enolates and formate reduction of allylic carbonates for polypropionate systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reactant involved in the synthesis of substrates for tin tetrachloride mediated fragmentation to produce ynones and ynoates, tandem oxidation coupling reactions with diamines, asymmetric allylic alkylation of enolates and formate reduction of allylic carbonates for polypropionate systems.

Solubility
Partly miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect from electrostatic charges. Ensure adequate ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30282-14-5
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00151157
InChI Key ODZTXUXIYGJLMC-UHFFFAOYSA-N
Synonym 2-hydroxycyclohexanone, adipoin, 2-hydroxy-1-cyclohexanone, cyclohexanone, 2-hydroxy, 2-hydroxy-cyclohexanone, cyclohexanone, 2-hydroxy-, 2r, cyclohexanone, 2-hydroxy-, dimer, cyclohexanone, dimer, zlchem 1128, acmc-20n5uh
PubChem CID 10785
ChEBI CHEBI:17878
IUPAC Name 2-hydroxycyclohexan-1-one
SMILES C1CCC(=O)C(C1)O

Specifications

Melting Point 100°C to 107°C
Boiling Point 83°C to 86°C (13 mmHg)
Flash Point 79°C (174°F)
Quantity 5 g
UN Number UN1325
Solubility Information Partly miscible in water.
Formula Weight 228.28
Percent Purity ≥90%
Chemical Name or Material 2-Hydroxycyclohexanone dimer

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.