missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxycinnamic Acid, TRC

Product Code. 30398351
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30398351 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398351 Supplier Toronto Research Chemicals Supplier No. TRCH91832310G
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Coumaric acid
CAS 614-60-8
Molecular Formula C9 H8 O3
Quantity 10 g
Synonym 2-Propenoic acid, 3-(2-hydroxyphenyl)-, (2E)-, 2-Propenoic acid, 3-(2-hydroxyphenyl)-, (E)-, Cinnamic acid, o-hydroxy-, (E)- (8CI), (2E)-3-(2-Hydroxyphenyl)-2-propenoic acid, (E)-2-Hydroxycinnamic acid, (E)-3-(2-Hydroxyphenyl)acrylic acid, (E)-o-Hydroxycinnamic acid, NSC 32952, o-Hydroxy-trans-cinnamic acid, trans-2-Hydroxycinnamic acid, trans-o-Coumaric acid, trans-o-Hydroxycinnamic acid
InChI Formula InChI=1S/C9H8O3/c10-8-4-2-1-3-7(8)5-6-9(11)12/h1-6,10H,(H,11,12)/b6-5+
SMILES OC(=O)\C=C\c1ccccc1O
IUPAC Name (E)-3-(2-hydroxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol) 164.16
Formula Weight 164.0473
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.