missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxycinnamaldehyde, TRC

Product Code. 30391074
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30391074 500 mg
30408647 1 g
30393319 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391074 Supplier Toronto Research Chemicals Supplier No. TRCH918325500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60125-23-7
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
Synonym (E)-3-(2-Hydroxyphenyl)-2-propenal, (2E)-3-(2-Hydroxyphenyl)-2-propenal, (E)-3-(2-Hydroxyphenyl)acrylaldehyde, o-Hydroxy-trans-cinnamaldehyde, trans-2-Hydroxycinnamaldehyde
IUPAC Name (E)-3-(2-hydroxyphenyl)prop-2-enal
SMILES Oc1ccccc1\C=C\C=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H8O2/c10-7-3-5-8-4-1-2-6-9(8)11/h1-7,11H/b5-3+
Formula Weight 148.0524
Chemical Name or Material 2-Hydroxycinnamaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.