missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxycaprylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Hydroxycaprylic Acid |
| CAS | 617-73-2 |
| Molecular Formula | C8H16O3 |
| Quantity | 50 mg |
| Synonym | 2-Hydroxycaprylic Acid, Octanoic acid, 2-hydroxy-, 2-Hydroxyoctanoic acid, (±)-2-Hydroxyoctanoic acid, (±)-α-Hydroxycaprylic acid, 2-Hydroxycaprylic acid, DL-2-Hydroxycaprylic acid, DL-2-Hydroxyoctanoic acid, NSC 189703, α-Hydroxy-n-caprylic acid, α-Hydroxycaprylic acid, α-Hydroxyoctanoic acid, 2-Hydroxycaprylic Acid, Octanoic acid, 2-hydroxy-, 2-Hydroxyoctanoic acid, (±)-2-Hydroxyoctanoic acid, (±)-α-Hydroxycaprylic acid, 2-Hydroxycaprylic acid, DL-2-Hydroxycaprylic acid, DL-2-Hydroxyoctanoic acid, NSC 189703, α-Hydroxy-n-caprylic acid, α-Hydroxycaprylic acid, α-Hydroxyoctanoic acid |
| InChI Formula | InChI=1S/C8H16O3/c1-2-3-4-5-6-7(9)8(10)11/h7,9H,2-6H2,1H3,(H,10,11) |
| SMILES | CCCCCCC(O)C(=O)O |
| IUPAC Name | 2-hydroxyoctanoic acid |
| Molecular Weight (g/mol) | 160.21 |
| Formula Weight | 160.1099 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?