missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybicyclo[3.2.1]octane-6-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10750164
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10750164 1 g Amber Glass Bottle 1g
10209282 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10750164 Supplier Thermo Scientific Maybridge Supplier No. BTBG00035DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 257932-29-9
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.21
MDL Number MFCD00180256
InChI Key FSQSHXLJRFYYMV-UHFFFAOYNA-N
Synonym 2-hydroxybicyclo 3.2.1 octane-6-carboxylic acid, maybridge1_002398, 4-hydroxybicyclo 3.2.1 octane-7-carboxylic acid, bicyclo 3.2.1 octane-6-carboxylicacid, 2-hydroxy
PubChem CID 2802922
IUPAC Name 4-hydroxybicyclo[3.2.1]octane-7-carboxylic acid
SMILES OC1CCC2CC1CC2C(O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 170.21
Percent Purity 97%
Chemical Name or Material 2-Hydroxybicyclo[3.2.1]octane-6-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.