missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzonitrile, TRC

Product Code. 30402522
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Product Code. Quantity
30402522 5 g
30391165 25 g
30401738 100 g
3 options
This item is not returnable. View return policy

Product Code. 30402522

Brand: Toronto Research Chemicals TRCH8084805G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-20-1
Formula Weight 119.0371
InChI Formula InChI=1S/C7H5NO/c8-5-6-3-1-2-4-7(6)9/h1-4,9H
IUPAC Name 2-hydroxybenzonitrile
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.12
SMILES Oc1ccccc1C#N
Synonym 2-Cyanophenol, 2-Hydroxybenzonitrile, NSC 53558, Salicylnitrile, o-Cyanophenol, o-Hydroxybenzonitrile
Chemical Name or Material 2-Hydroxybenzonitrile

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.