missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzonitrile, 98%

Product Code. 11410164
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410164 5 g 5g
11420164 25 g 25g
11430164 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410164 Supplier Thermo Scientific Alfa Aesar Supplier No. A13606.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 611-20-1 | C7H5NO | 119.123 g/mol

2-Hydroxybenzonitrile is used as starting reagent during the synthesis of mono-alkoxyphenyloxazoline, furanoside.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxybenzonitrile is used as starting reagent during the synthesis of mono-alkoxyphenyloxazoline, furanoside.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-20-1
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.123
MDL Number MFCD00002145
InChI Key CHZCERSEMVWNHL-UHFFFAOYSA-N
Synonym 2-cyanophenol, o-cyanophenol, salicylonitrile, o-hydroxybenzonitrile, benzonitrile, 2-hydroxy, salicylnitrile, benzonitrile, hydroxy, benzonitrile, o-hydroxy, o-hydoxybenzonitrile, 2-hydroxy-benzonitrile
PubChem CID 11907
IUPAC Name 2-hydroxybenzonitrile
SMILES C1=CC=C(C(=C1)C#N)O

Specifications

Melting Point 92°C to 95°C
Boiling Point 149°C (14 mmHg)
Quantity 5 g
Beilstein 1210029
Solubility Information Soluble in water.
Formula Weight 119.12
Percent Purity 98%
Chemical Name or Material 2-Hydroxybenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.