missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyamino-3-methyl-3H-imidazo[4,5-f]quinoline, TRC

Product Code. 30398870
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398870 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398870 Supplier Toronto Research Chemicals Supplier No. TRCH78500010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxyamino-3-methyl-3H-imidazo[4,5-f]quinoline
CAS 77314-23-9
Molecular Formula C11 H10 N4 O
Quantity 10 mg
InChI Formula InChI=1S/C11H10N4O/c1-15-9-5-4-8-7(3-2-6-12-8)10(9)13-11(15)14-16/h2-6,16H,1H3,(H,13,14)
SMILES Cn1c(NO)nc2c3cccnc3ccc12
IUPAC Name N-(3-methylimidazo[4,5-f]quinolin-2-yl)hydroxylamine
Molecular Weight (g/mol) 214.22
Formula Weight 214.0855
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.