missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyalmotriptan, TRC

Product Code. 30397594
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30397594 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397594 Supplier Toronto Research Chemicals Supplier No. TRCH74998510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1309457-19-9
Molecular Formula C17H25N3O3S
Quantity 10 mg
Synonym 3-[2-(Dimethylamino)ethyl]-5-[(1-pyrrolidinylsulfonyl)methyl]-1H-Indol-2-ol, Almotriptan 2-Hydroxy Impurity
InChI Formula InChI=1S/C17H25N3O3S/c1-19(2)10-7-14-15-11-13(5-6-16(15)18-17(14)21)12-24(22,23)20-8-3-4-9-20/h5-6,11,18,21H,3-4,7-10,12H2,1-2H3
SMILES OC(N1)=C(CCN(C)C)C2=C1C=CC(CS(=O)(N3CCCC3)=O)=C2
Molecular Weight (g/mol) 351.46
Formula Weight 351.1617
Chemical Name or Material 2-Hydroxyalmotriptan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.