missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxyacetophenone, MedChemExpress

Product Code. 16439426
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16439426 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16439426 Supplier Medchem Express Supplier No. HYY1426
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 2'-Hydroxyacetophenone is found in alcoholic beverages. 2'-Hydroxyacetophenone is present in tomato, cassia, fried beef, rum, whiskey, cocoa, coffee and black tea. 2'-Hydroxyacetophenone is a flavouring ingredient. Building block in chemical synthesis.

2'-Hydroxyacetophenone is found in alcoholic beverages. 2'-Hydroxyacetophenone is present in tomato, cassia, fried beef, rum, whiskey, cocoa, coffee and black tea. 2'-Hydroxyacetophenone is a flavouring ingredient. Building block in chemical synthesis.

Specifications

Chemical Name or Material 2'-Hydroxyacetophenone
CAS 118-93-4
Color Colorless to Light Yellow
Purity Grade Notes Research
Molecular Formula C8H8O2
Quantity 100 mg
Synonym o-Hydroxyacetophenone o-Acetylphenol
Solubility Information DMSO : 100 mg/mL (734.48 mM; Need ultrasonic)
SMILES CC(C1=CC=CC=C1O)=O
Molecular Weight (g/mol) 136.15
Formula Weight 136.15
Percent Purity 97.0%
Grade Research
Physical Form Liquid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.