missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxyacetophenone, 98%

Product Code. 11444244
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444244 100 g 100g
11454244 500 g 500g
11464244 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444244 Supplier Thermo Scientific Alfa Aesar Supplier No. A12550.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 118-93-4 | C8H8O2 | 136.15 g/mol

2'-Hydroxyacetophenone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Hydroxyacetophenone is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-93-4
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002219
InChI Key JECYUBVRTQDVAT-UHFFFAOYSA-N
Synonym 2'-hydroxyacetophenone, 2-acetylphenol, 1-2-hydroxyphenyl ethanone, o-hydroxyacetophenone, o-acetylphenol, o-hydroxyphenyl methyl ketone, ethanone, 1-2-hydroxyphenyl, 1-2-hydroxyphenyl ethan-1-one, acetophenone, 2'-hydroxy, 2-hydroxyphenyl methyl ketone
PubChem CID 8375
IUPAC Name 1-(2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1O

Specifications

Melting Point 3°C to 6°C
Density 1.13
Boiling Point 218°C
Flash Point 106°C (222°F)
Refractive Index 1.5584
Quantity 100 g
Beilstein 386123
Solubility Information Slightly soluble in water.
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material 2'-Hydroxyacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 2'-Hydroxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2'-Hydroxyacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.