missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-propoxycarbazone, TRC

Product Code. 30011815
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011815 100 mg 100mg
30005580 25 mg 25mg
30011561 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011815 Supplier Toronto Research Chemicals Supplier No. TRCH185620100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496925-02-1
Molecular Formula C15 H18 N4 O8 S
Molecular Weight (g/mol) 414.39
Synonym Benzoic acid, 2-[[[[4,5-dihydro-3-(2-hydroxypropoxy)-4-methyl-5-oxo-1H-1,2,4-triazol-1-yl]carbonyl]amino]sulfonyl]-, methyl ester
IUPAC Name methyl 2-[[3-(2-hydroxypropoxy)-4-methyl-5-oxo-1,2,4-triazole-1-carbonyl]sulfamoyl]benzoate
SMILES COC(=O)c1ccccc1S(=O)(=O)NC(=O)N2N=C(OCC(C)O)N(C)C2=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H18N4O8S/c1-9(20)8-27-14-16-19(15(23)18(14)2)13(22)17-28(24,25)11-7-5-4-6-10(11)12(21)26-3/h4-7,9,20H,8H2,1-3H3,(H,17,22)
Formula Weight 414.0845
Percent Purity >95%
Chemical Name or Material Propoxycarbazone-2-hydroxypropoxy
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.