missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy Oleic Acid, TRC

Product Code. 30399048
Change view
Click to view available options
Quantity:
5 mg
25 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399048 25 mg
30400851 5 mg
30391880 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399048 Supplier Toronto Research Chemicals Supplier No. TRCH94876525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56472-29-8
Molecular Formula C18H34O3
Molecular Weight (g/mol) 298.46
Synonym (9Z)-2-Hydroxy-9-octadecenoic Acid, (Z)-2-Hydroxyoctadec-9-enoic acid, 2-Hydroxyoleic Acid, Minerval, cis-2-Hydroxy-9-octadecenoic Acid, α-Hydroxyoleic Acid
SMILES OC(CCCCCC/C=C\CCCCCCCC)C(O)=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C18H34O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(19)18(20)21/h9-10,17,19H,2-8,11-16H2,1H3,(H,20,21)/b10-9-
Formula Weight 298.2508
Chemical Name or Material 2-Hydroxy Oleic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.