missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-N-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]acetamide, TRC

Product Code. 30390175
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390175 1 mg -
30407119 5 mg 5mg
30389306 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390175 Supplier Toronto Research Chemicals Supplier No. TRCH9181201MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 914465-63-7
Molecular Formula C16 H15 N5 O2
Molecular Weight (g/mol) 309.32
Synonym Acetamide, 2-hydroxy-N-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, Acetamide, 2-hydroxy-N-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]- (9CI)
IUPAC Name 2-hydroxy-N-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]acetamide
SMILES OCC(=O)NCc1ccc(cc1)c2ccccc2c3nnn[nH]3

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C16H15N5O2/c22-10-15(23)17-9-11-5-7-12(8-6-11)13-3-1-2-4-14(13)16-18-20-21-19-16/h1-8,22H,9-10H2,(H,17,23)(H,18,19,20,21)
Formula Weight 309.1226
Chemical Name or Material 2-Hydroxy-N-((2'-(2H-tetrazol-5-yl)(1,1'-biphenyl)-4-yl)methyl)acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.