missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy L-Tryptophan Hydrochloride, TRC

Product Code. 30010665
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010665 250 mg 250mg
30010405 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010665 Supplier Toronto Research Chemicals Supplier No. TRCH975680250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 881025-90-7
Molecular Formula C11H13ClN2O3
Molecular Weight (g/mol) 256.69
IUPAC Name (2S)-2-amino-3-(2-hydroxy-1H-indol-3-yl)propanoic acid
SMILES N[C@H](C(O)=O)CC1=C(O)NC2=CC=CC=C21.Cl

Specifications

InChI Formula InChI=1S/C11H12N2O3/c12-8(11(15)16)5-7-6-3-1-2-4-9(6)13-10(7)14/h1-4,8,13-14H,5,12H2,(H,15,16)/t8-/m0/s1
Quantity 250 mg
Formula Weight 220.0848
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material 2-Hydroxy L-Tryptophan Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.