missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy Hippuric Acid, TRC

Product Code. 30397517
Change view
Click to view available options
Quantity:
250 mg
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30397517 25 g
30389665 250 mg
30404774 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397517 Supplier Toronto Research Chemicals Supplier No. TRCH94288525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 487-54-7
Molecular Formula C9 H9 N O4
Molecular Weight (g/mol) 195.17
Synonym Glycine, N-(2-hydroxybenzoyl)-Hippuric acid, o-hydroxy- (8CI), Salicyluric acid (6CI), (2-Hydroxybenzoyl)glycine, 2-Hydroxyhippuric acid, N-(o-Hydroxybenzoyl)glycine, N-Salicyloylglycine, NSC 524135, Salicyloylglycine, o-Hydroxyhippuric acid
IUPAC Name 2-[(2-hydroxybenzoyl)amino]acetic acid
SMILES OC(=O)CNC(=O)c1ccccc1O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C9H9NO4/c11-7-4-2-1-3-6(7)9(14)10-5-8(12)13/h1-4,11H,5H2,(H,10,14)(H,12,13)
Formula Weight 195.0532
Chemical Name or Material 2-Hydroxy Hippuric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.