missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-estrone-6-N3-adenine (90%), TRC

Product Code. 30402796
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402796 10 mg
30408445 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402796 Supplier Toronto Research Chemicals Supplier No. TRCH94216510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 912671-65-9
Molecular Formula C23H25N5O3
Molecular Weight (g/mol) 419.48
Synonym 6-(6-Amino-3H-purin-3-yl)-2,3-dihydroxy-estra-1,3,5(10)-trien-17-one
SMILES OC1=C(O)C=C2C(C(N3C4=NC=NC4=C(N)N=C3)C[C@]5([H])[C@]2([H])CC[C@@]6(C)[C@@]5([H])CCC6=O)=C1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H25N5O3/c1-23-5-4-11-12-7-17(29)18(30)8-14(12)16(6-13(11)15(23)2-3-19(23)31)28-10-27-21(24)20-22(28)26-9-25-20/h7-11,13,15-16,24,29-30H,2-6H2,1H3,(H,25,26)/t11-,13-,15+,16?,23+/m1/s1
Formula Weight 419.1957
Chemical Name or Material 2-Hydroxy-estrone-6-N3-adenine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.