missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy Estrone 6-N3-Adenine 2,3-O-Di-t-Butyloxycarbonyl, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Hydroxy Estrone 6-N3-Adenine 2,3-O-Di-t-Butyloxycarbonyl |
| Analyte or Component Names | 2-Hydroxy Estrone 6-N3-Adenine 2,3-O-Di-t-Butyloxycarbonyl |
| Shipping Condition | Room Temperature |
| Molecular Formula | C33 H41 N5 O7 |
| Synonym | (8R,9S,13S,14S)-6-(6-Amino-3H-purin-3-yl)-13-methyl-17-oxo-7,8,9,11,12,13,14,15,16,17-decahydro-6H-cyclopenta[a]phenanthrene-2,3-diyl Di-t-Butyl Dicarbonate |
| InChI Formula | InChI=1S/C33H41N5O7/c1-31(2,3)44-29(40)42-23-13-18-17-10-11-33(7)21(8-9-25(33)39)19(17)12-22(38-16-37-27(34)26-28(38)36-15-35-26)20(18)14-24(23)43-30(41)45-32(4,5)6/h13-17,19,21-22H,8-12,34H2,1-7H3/t17-,19-,21+,22?,33+/m1/s1 |
| Solution Type | Neat |
| SMILES | CC(C)(C)OC(=O)Oc1cc2[C@H]3CC[C@@]4(C)[C@@H](CCC4=O)[C@@H]3CC(c2cc1OC(=O)OC(C)(C)C)n5cnc(N)c6ncnc56 |
| IUPAC Name | [(8R,9S,13S,14S)-6-(6-aminopurin-3-yl)-13-methyl-2-[(2-methylpropan-2-yl)oxycarbonyloxy]-17-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-yl] tert-butyl carbonate |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?