missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy Desipramine-d3, TRC

Product Code. 30399592
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399592 10 mg
30392422 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30399592 Supplier Toronto Research Chemicals Supplier No. TRCH93650210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189998-63-7
Molecular Formula C18H19D3N2O
Molecular Weight (g/mol) 285.4
Synonym 10,11-Dihydro-5-[3-(methyl-d3-amino)propyl]-5H-dibenz[b,f]azepin-2-ol, 2-Hydroxydemethylimipramine-d3, 2-Hydroxydesmethylimipramine-d3, GP 36329-d3
IUPAC Name 11-[3-(trideuteriomethylamino)propyl]-5,6-dihydrobenzo[b][1]benzazepin-3-ol
SMILES [2H]C([2H])([2H])NCCCN1c2ccccc2CCc3cc(O)ccc13

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H22N2O/c1-19-11-4-12-20-17-6-3-2-5-14(17)7-8-15-13-16(21)9-10-18(15)20/h2-3,5-6,9-10,13,19,21H,4,7-8,11-12H2,1H3/i1D3
Formula Weight 285.192
Chemical Name or Material 2-Hydroxy Desipramine-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.