missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy Desipramine Beta-D-Glucuronide, TRC

Product Code. 30394220
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394220 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394220 Supplier Toronto Research Chemicals Supplier No. TRCH9365101MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxy Desipramine beta-D-Glucuronide
CAS 25521-31-7
Molecular Formula C24 H30 N2 O7
Quantity 1 mg
Synonym 2-Hydroxy Desipramine-B-D-Glucuronide
InChI Formula InChI=1S/C24H30N2O7/c1-25-11-4-12-26-17-6-3-2-5-14(17)7-8-15-13-16(9-10-18(15)26)32-24-21(29)19(27)20(28)22(33-24)23(30)31/h2-3,5-6,9-10,13,19-22,24-25,27-29H,4,7-8,11-12H2,1H3,(H,30,31)/t19-,20-,21+,22-,24?/m0/s1
SMILES CNCCCN1c2ccccc2CCc3cc(OC4O[C@@H]([C@@H](O)[C@H](O)[C@H]4O)C(=O)O)ccc13
IUPAC Name (2S,3S,4S,5R)-3,4,5-trihydroxy-6-[[11-[3-(methylamino)propyl]-5,6-dihydrobenzo[b][1]benzazepin-3-yl]oxy]oxane-2-carboxylic acid
Molecular Weight (g/mol) 458.5
Formula Weight 458.2053
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.