missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-bexarotene, TRC

Product Code. 30392231
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392231 50 mg
30403351 25 mg
30406304 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392231 Supplier Toronto Research Chemicals Supplier No. TRCH21221050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2122184-62-5
Molecular Formula C24H30O3
Molecular Weight (g/mol) 366.49
Synonym Bexarotene Impurity, 4-[2-Hydroxy-1-(5,6,7,8-tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)ethyl]benzoic Acid
IUPAC Name 4-[2-hydroxy-1-(3,5,5,8,8-pentamethyl-6,7-dihydronaphthalen-2-yl)ethyl]benzoic acid
SMILES Cc1cc2c(cc1C(CO)c3ccc(cc3)C(=O)O)C(C)(C)CCC2(C)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C24H30O3/c1-15-12-20-21(24(4,5)11-10-23(20,2)3)13-18(15)19(14-25)16-6-8-17(9-7-16)22(26)27/h6-9,12-13,19,25H,10-11,14H2,1-5H3,(H,26,27)
Formula Weight 366.2195
Chemical Name or Material 2-Hydroxy-bexarotene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.