missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-9H-thioxanthen-9-one, TRC

Product Code. 30394355
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394355 1 g -
30407281 500 mg -
30403603 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394355 Supplier Toronto Research Chemicals Supplier No. TRCH9560831G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31696-67-0
Molecular Formula C13H8O2S
Molecular Weight (g/mol) 228.27
Synonym 2-Hydroxythioxanthen-9-one, 2-Hydroxythioxanthone
IUPAC Name 2-hydroxythioxanthen-9-one
SMILES Oc1ccc2Sc3ccccc3C(=O)c2c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H8O2S/c14-8-5-6-12-10(7-8)13(15)9-3-1-2-4-11(9)16-12/h1-7,14H
Formula Weight 228.0245
Chemical Name or Material 2-Hydroxy-9H-thioxanthen-9-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.