missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-6-methoxybenzaldehyde, 98+%

Product Code. 11317936
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317936 5 g 5g
11307936 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317936 Supplier Thermo Scientific Alfa Aesar Supplier No. L13145.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 700-44-7 | C8H8O3 | 152.149 g/mol

2-Hydroxy-6-methoxybenzaldehyde is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-6-methoxybenzaldehyde is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 700-44-7
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00151830
InChI Key DZJPDDVDKXHRLF-UHFFFAOYSA-N
PubChem CID 69692
IUPAC Name 2-hydroxy-6-methoxybenzaldehyde
SMILES COC1=CC=CC(=C1C=O)O

Specifications

Melting Point 68°C to 74°C
Quantity 5 g
Beilstein 1101524
Sensitivity Air Sensitive
Synonym 6-methoxysalicylaldehyde, 2-hydroxy-6-methoxy-benzaldehyde, 2-hydroxy-6-methoxybenzal dehyde, 2-hydroxy-6-methoxy benzaldehyde, benzaldehyde, 2-hydroxy-6-methoxy, pubchem8210, 2-methoxy-6-hydroxybenzaldehyde, benzaldehyde,2-hydroxy-6-methoxy, 2-hydroxy-6-methyloxy benzaldehyde
Solubility Information Slightly soluble in water.
Formula Weight 152.15
Percent Purity ≥98%
Chemical Name or Material 2-Hydroxy-6-methoxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.