missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-6-aminopurine, 98%

Product Code. 10185000 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass Bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10185000 250 mg Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10185000 Supplier Thermo Scientific Acros Supplier No. 241752500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3373-53-3 | C5H5N5O | 151.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3373-53-3
CAS Min % 97.5
CAS Max % 100.0
Melting Point 300°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C5H5N5O
MDL Number MFCD00142931
Quantity 250 mg
Synonym isoguanine, 2-hydroxyadenine, 2-hydroxy-6-aminopurine, 6-amino-3,7-dihydro-2h-purin-2-one, 6-amino-2-hydroxypurine, 6-amino-7h-purin-2-ol, 2-oxoadenine, 6-amino-1,3-dihydro-2h-purin-2-one, 2-oxyadenine, 6-amino-9h-purin-2-ol
Solubility Information Solubility in water: .. Other solubilities: soluble in alkali,practically insoluble in most organic solvents
InChI Key DRAVOWXCEBXPTN-UHFFFAOYSA-N
SMILES NC1=C2NC=NC2=NC(=O)N1
IUPAC Name 6-amino-1,7-dihydropurin-2-one
Molecular Weight (g/mol) 151.13
PubChem CID 76900
ChEBI CHEBI:62462
Formula Weight 151.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Hydroxy-6-aminopurine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.