missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Product Code. 10143304 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10143304 1 g Glass bottle 1g
10103354 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10143304 Supplier Thermo Scientific Acros Supplier No. 247260010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 33252-63-0
Molecular Formula C6H4F3NO
Molecular Weight (g/mol) 163.10
MDL Number MFCD00042315
InChI Key BYRJSCNPUHYZQE-UHFFFAOYSA-N
Synonym 2-hydroxy-5-trifluoromethylpyridine, 5-trifluoromethyl pyridin-2-ol, 2-hydroxy-5-trifluoromethyl pyridine, 5-trifluoromethyl-2-pyridinol, 5-trifluoromethyl-pyridin-2-ol, 5-trifluoromethyl-2-pyridone, unii-1ccp81h07b, 5-trifluoromethyl pyridin-2 1h-one, 5-trifluoromethyl-1,2-dihydropyridin-2-one, 5-trifloromethyl-2-1h-pyridinone
PubChem CID 147443
IUPAC Name 5-(trifluoromethyl)-1H-pyridin-2-one
SMILES FC(F)(F)C1=CNC(=O)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 145.0°C to 149.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 163.1
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-5-(trifluoromethyl)pyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxy-5-(trifluoromethyl)pyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.