missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Hydroxy-5-(trifluoromethoxy)benzonitrile, 95%

Product Code. 15487545
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15487545

Brand: Alfa Aesar™ H26101.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 875664-40-7
Molecular Formula C8H4F3NO2
Molecular Weight (g/mol) 203.12
MDL Number MFCD04973781
InChI Key GOAVRYAOBXORSY-UHFFFAOYSA-N
Synonym 2-hydroxy-5-trifluoromethoxy benzonitrile
PubChem CID 24721643
IUPAC Name 2-hydroxy-5-(trifluoromethoxy)benzonitrile
SMILES C1=CC(=C(C=C1OC(F)(F)F)C#N)O

Specifications

Melting Point 84°C to 90°C
Quantity 250 mg
UN Number UN3439
Formula Weight 203.12
Percent Purity 95%
Chemical Name or Material 2-Hydroxy-5-(trifluoromethoxy)benzonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.