missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-(trifluoromethoxy)benzaldehyde, 99%

Product Code. 10536114 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10536114 1 g Glass bottle 1g
10011614 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10536114 Supplier Thermo Scientific Acros Supplier No. 398200010
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 93249-62-8 | C8H5F3O3 | 206.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93249-62-8
Molecular Formula C8H5F3O3
Molecular Weight (g/mol) 206.12
MDL Number MFCD00075249
InChI Key WQUZBERVMUEJTD-UHFFFAOYSA-N
PubChem CID 1268058
IUPAC Name 2-hydroxy-5-(trifluoromethoxy)benzaldehyde
SMILES C1=CC(=C(C=C1OC(F)(F)F)C=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 31.0°C to 33.0°C
Color White to Yellow
Boiling Point 82.0°C (60.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CF3OC6H3(OH)CHO
Quantity 1 g
Synonym 2-hydroxy-5-trifluoromethoxy benzaldehyde, 5-trifluoromethoxy salicylaldehyde, benzaldehyde, 2-hydroxy-5-trifluoromethoxy, acmc-20antc, 5-trifluoromethoxysalicylaldehyde, 5-trifluoromethoxy-salicylaldehyde, 2-hydroxy-5-trifluoromethoxy-benzaldehyde, 20hydroxy-5-trifluoromethoxy benzaldehyde, 2-oxidanyl-5-trifluoromethyloxy benzaldehyde, benzaldehyde,2-hydroxy-5-trifluoromethoxy
Formula Weight 206.12
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material 2-Hydroxy-5-(trifluoromethoxy)benzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxy-5-(trifluoromethoxy)benzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.