missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-(phenyldiazenyl)benzoic Acid (Phenylazosalicylic Acid), Mikromol™

Product Code. 17923966
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17923966 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17923966 Supplier Mikromol Supplier No. MM0217.12
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Hydroxy-5-(phenyldiazenyl)benzoic Acid (Phenylazosalicylic Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxy-5-(phenyldiazenyl)benzoic Acid (Phenylazosalicylic Acid)
CAS 3147-53-3
Analyte or Component Names 2-Hydroxy-5-(phenyldiazenyl)benzoic Acid (Phenylazosalicylic Acid)
Shipping Condition Room Temperature
Molecular Formula C13 H10 N2 O3
Certifications/Compliance ISO 9001
Synonym 2-Hydroxy-5-(phenyldiazenyl)benzoic Acid, Mesalazine Imp. I (EP), Phenylazosalicylic Acid, Mesalazine Impurity I
InChI Formula InChI=1S/C13H10N2O3/c16-12-7-6-10(8-11(12)13(17)18)15-14-9-4-2-1-3-5-9/h1-8,16H,(H,17,18)/b15-14+
SMILES OC(=O)c1cc(ccc1O)N=Nc2ccccc2
IUPAC Name 2-hydroxy-5-[(E)-phenyldiazenyl]benzoic acid
Molecular Weight (g/mol) 242.23
Quantity 50 mg
Formula Weight 242.0691 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.