missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-octanoylbenzoic Acid, TRC

Product Code. 30405019
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30405019 100 mg -
30403673 10 mg 10mg
30396253 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30405019 Supplier Toronto Research Chemicals Supplier No. TRCH951198100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78418-01-6
Molecular Formula C15H20O4
Molecular Weight (g/mol) 264.32
Synonym Benzoic acid, 2-hydroxy-5-(1-oxooctyl)- (9CI, ACI), 2-Hydroxy-5-(1-oxooctyl)benzoic acid (ACI), 2-Hydroxy-5-octanoylbenzoic acid, 5-n-Octanoylsalicylic acid, 5-Octanoylsalicylic acid, Effaclark, n-Octanoyl-5-salicylic acid, β-Lipohydroxy acid
IUPAC Name 2-hydroxy-5-octanoylbenzoic acid
SMILES CCCCCCCC(=O)c1ccc(O)c(c1)C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H20O4/c1-2-3-4-5-6-7-13(16)11-8-9-14(17)12(10-11)15(18)19/h8-10,17H,2-7H2,1H3,(H,18,19)
Formula Weight 264.1362
Chemical Name or Material 2-Hydroxy-5-octanoylbenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.