missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-nitropyridine, 99+%, Thermo Scientific™

Product Code. 10326520 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10326520 10 g Glass bottle 10g
10172440 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10326520 Supplier Thermo Scientific Acros Supplier No. 121680100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 5418-51-9
Molecular Formula C5H4N2O3
Molecular Weight (g/mol) 140.1
MDL Number MFCD00006276
InChI Key XKWSQIMYNVLGBO-UHFFFAOYSA-N
Synonym 2-hydroxy-5-nitropyridine, 5-nitropyridin-2-ol, 5-nitro-2-pyridinol, 2 1h-pyridinone, 5-nitro, 5-nitropyridin-2 1h-one, 5-nitro-2-hydroxypyridine, 5-nitro-2-pyridone, 5-nitro-1,2-dihydropyridin-2-one, 5-nitro-2-pyridol, pubchem1205
PubChem CID 79453
IUPAC Name 5-nitro-1H-pyridin-2-one
SMILES C1=CC(=O)NC=C1[N+](=O)[O-]

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 188°C to 191°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Linear Formula HO(C5H3N)NO2
Quantity 10 g
Beilstein 21, I, 202
Solubility Information Solubility in water: .. Other solubilities: soluble in alkalies,slightly soluble in organic solvents
Formula Weight 140.1
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-5-nitropyridine, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxy-5-nitropyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.