Learn More
2-Hydroxy-5-nitropyridine, 98+%, Thermo Scientific Chemicals
£33.60 - £93.50
Chemical Identifiers
CAS | 5418-51-9 |
---|---|
Molecular Formula | C5H4N2O3 |
Molecular Weight (g/mol) | 140.098 |
MDL Number | MFCD00006276 |
InChI Key | XKWSQIMYNVLGBO-UHFFFAOYSA-N |
Synonym | 2-hydroxy-5-nitropyridine, 5-nitropyridin-2-ol, 5-nitro-2-pyridinol, 2 1h-pyridinone, 5-nitro, 5-nitropyridin-2 1h-one, 5-nitro-2-hydroxypyridine, 5-nitro-2-pyridone, 5-nitro-1,2-dihydropyridin-2-one, 5-nitro-2-pyridol, pubchem1205 |
PubChem CID | 79453 |
IUPAC Name | 5-nitro-1H-pyridin-2-one |
SMILES | C1=CC(=O)NC=C1[N+](=O)[O-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11461638
|
Thermo Scientific Alfa Aesar
A13921.14 |
25 g |
£33.60
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11471638
|
Thermo Scientific Alfa Aesar
A13921.22 |
100 g |
£93.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A comparison of catalysts promotes imidazolide couplings including the identification of 2-hydroxy-5-nitropyridine as a new, safe, and effective catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA comparison of catalysts promotes imidazolide couplings including the identification of 2-hydroxy-5-nitropyridine as a new, safe, and effective catalyst.
Solubility
Soluble in hot water and alkali liquor, Insoluble in most of organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
5418-51-9 | |
140.098 | |
XKWSQIMYNVLGBO-UHFFFAOYSA-N | |
79453 | |
C1=CC(=O)NC=C1[N+](=O)[O-] |
C5H4N2O3 | |
MFCD00006276 | |
2-hydroxy-5-nitropyridine, 5-nitropyridin-2-ol, 5-nitro-2-pyridinol, 2 1h-pyridinone, 5-nitro, 5-nitropyridin-2 1h-one, 5-nitro-2-hydroxypyridine, 5-nitro-2-pyridone, 5-nitro-1,2-dihydropyridin-2-one, 5-nitro-2-pyridol, pubchem1205 | |
5-nitro-1H-pyridin-2-one |
Specifications
5418-51-9 | |
C5H4N2O3 | |
120352 | |
Soluble in hot water and alkali liquor,Insoluble in most of organic solvents. | |
C1=CC(=O)NC=C1[N+](=O)[O-] | |
140.098 | |
140.1 | |
2-Hydroxy-5-nitropyridine |
188°C to 191°C | |
MFCD00006276 | |
2-hydroxy-5-nitropyridine, 5-nitropyridin-2-ol, 5-nitro-2-pyridinol, 2 1h-pyridinone, 5-nitro, 5-nitropyridin-2 1h-one, 5-nitro-2-hydroxypyridine, 5-nitro-2-pyridone, 5-nitro-1,2-dihydropyridin-2-one, 5-nitro-2-pyridol, pubchem1205 | |
XKWSQIMYNVLGBO-UHFFFAOYSA-N | |
5-nitro-1H-pyridin-2-one | |
79453 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 226-525-7
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only