missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-nitrobenzaldehyde, 98%

Product Code. p-7080648
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11423983

Brand: Thermo Scientific Alfa Aesar A10264.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 97-51-8 | C7H5NO4 | 167.12 g/mol

2-Hydroxy-5-nitrobenzaldehyde is used in the synthetic preparation of coumarin (C755380), an pharmaceutic aid that is found in tonka beans, lavender oil, woodruff, sweet clover.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-5-nitrobenzaldehyde is used in the synthetic preparation of coumarin (C755380), an pharmaceutic aid that is found in tonka beans, lavender oil, woodruff, sweet clover.

Solubility
Soluble in water (partly).

Notes
Air & Light Sensitive,. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-51-8
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007337
InChI Key IHFRMUGEILMHNU-UHFFFAOYSA-N
Synonym 5-nitrosalicylaldehyde, benzaldehyde, 2-hydroxy-5-nitro, salicylaldehyde, 5-nitro, 5-nitrosaliclaldehyde, 2-hydroxy-5-nitro-benzaldehyde, 5-nitro salicylaldehdye, timtec-bb sbb003885, labotest-bb lt00920994, salicylaldehyde, 5-nitro-6ci,7ci,8ci, pubchem2052
PubChem CID 66808
IUPAC Name 2-hydroxy-5-nitrobenzaldehyde
SMILES OC1=CC=C(C=C1C=O)[N+]([O-])=O

Specifications

Melting Point 123°C to 127°C
Quantity 5 g
Beilstein 512565
Sensitivity Air and light sensitive
Solubility Information Soluble in water (partly).
Formula Weight 167.12
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-5-nitrobenzaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.