missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-(1-hydroxyethyl)benzamide, TRC

Product Code. 30399813
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399813 100 mg 100mg
30388831 1 g 1g
30405748 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399813 Supplier Toronto Research Chemicals Supplier No. TRCH415055100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C9 H11 N O3
Molecular Weight (g/mol) 181.189
IUPAC Name 2-hydroxy-5-(1-hydroxyethyl)benzamide
SMILES CC(O)c1ccc(O)c(c1)C(=O)N

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H11NO3/c1-5(11)6-2-3-8(12)7(4-6)9(10)13/h2-5,11-12H,1H3,(H2,10,13)
Formula Weight 181.074
Chemical Name or Material 2-Hydroxy-5-(1-hydroxyethyl)benzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.