missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-(trifluoromethyl)pyrimidine, 99%, Thermo Scientific™

Product Code. 15426628
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15426628 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15426628 Supplier Thermo Scientific Alfa Aesar Supplier No. H60151.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104048-92-2
Molecular Formula C5H3F3N2O
Molecular Weight (g/mol) 164.09
MDL Number MFCD00192529
InChI Key WCEBFEVZTGLOHC-UHFFFAOYSA-N
Synonym 2-hydroxy-4-trifluoromethyl pyrimidine, 4-trifluoromethyl pyrimidin-2-ol, 4-trifluoromethyl pyrimidin-2 1h-one, 2-hydroxy-4-trifluoromethylpyrimidine, 4-trifluoromethyl-2-pyrimidinol, 6-trifluoromethyl-1h-pyrimidin-2-one, 2 1h-pyrimidinone, 4-trifluoromethyl, pubchem7201, enamine_002404, acmc-2098am
PubChem CID 1240050
SMILES FC(F)(F)C1=CC=NC(=O)N1

Specifications

Melting Point 223°C to 227°C
Quantity 1 g
IUPAC Name 6-(trifluoromethyl)-1H-pyrimidin-2-one
Formula Weight 164.09
Percent Purity 99%
Chemical Name or Material 2-Hydroxy-4-(trifluoromethyl)pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.