missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-(trifluoromethyl)pyridine, 98%, Thermo Scientific™

Product Code. 11329979
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11329979 1 g 1g
11319979 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329979 Supplier Thermo Scientific Alfa Aesar Supplier No. H25756.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Hydroxy-4-(trifluoromethyl)pyridine is used in the production of agrochemicals and pharmaceuticals.

Solubility
Soluble in methanol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50650-59-4
Molecular Formula C6H4F3NO
Molecular Weight (g/mol) 163.10
MDL Number MFCD00153195,MFCD09029339
InChI Key IKHLLNMSMFVTLP-UHFFFAOYSA-N
Synonym 2-hydroxy-4-trifluoromethyl pyridine, 4-trifluoromethyl pyridin-2-ol, 4-trifluoromethyl pyridin-2 1h-one, 4-trifluoromethyl-pyridin-2-ol, 4-trifluoromethyl-2-pyridone, 2-hydroxy-4-trifluoromethylpyridine, 4-trifluoromethyl-2 1h pyridinone, 4-trifluoromethyl-1h-pyridin-2-one, 2 1h-pyridinone, 4-trifluoromethyl, 4-trifluoromethyl-2 1h-pyridinone
PubChem CID 2775130
SMILES FC(F)(F)C1=CC(=O)NC=C1

Specifications

Melting Point 158°C to 160°C
Quantity 1 g
Solubility Information Soluble in methanol.
IUPAC Name 4-(trifluoromethyl)-1,2-dihydropyridin-2-one
Formula Weight 163.1
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-4-(trifluoromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.