missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-nitrobenzaldehyde, TRC

Product Code. 30393980
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393980 1 g -
30394527 10 g -
30404994 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393980 Supplier Toronto Research Chemicals Supplier No. TRCH9486401G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2460-58-4
Molecular Formula C7 H5 N O4
Molecular Weight (g/mol) 167.12
IUPAC Name 2-hydroxy-4-nitrobenzaldehyde
SMILES Oc1cc(ccc1C=O)[N+](=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H5NO4/c9-4-5-1-2-6(8(11)12)3-7(5)10/h1-4,10H
Formula Weight 167.0219
Chemical Name or Material 2-Hydroxy-4-nitro-benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.