missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methyl-5-nitropyridine, 98%, Thermo Scientific™

Product Code. 10617071 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10617071 1 g Glass bottle 1g
10760585 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10617071 Supplier Thermo Scientific Acros Supplier No. 319570010
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 21901-41-7
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.13
MDL Number MFCD00010690
InChI Key AIEHUZHKFUNHCJ-UHFFFAOYSA-N
Synonym 2-hydroxy-4-methyl-5-nitropyridine, 4-methyl-5-nitropyridin-2-ol, 2-hydroxy-5-nitro-4-picoline, 2-hydroxy-5-nitro-4-methyl pyridine, 2-hydroxy-5-nitro-4-methylpyridine, 4-methyl-5-nitropyridin-2 1h-one, 4-methyl-5-nitro-2-pyridinol, 4-methyl-5-nitro-1,2-dihydropyridin-2-one, 4-methyl-5-nitro-pyridin-2-ol, 2 1h-pyridinone, 4-methyl-5-nitro
PubChem CID 345371
IUPAC Name 4-methyl-5-nitro-1H-pyridin-2-one
SMILES CC1=CC(=O)NC=C1[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 186°C to 190°C
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 154.13
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-4-methyl-5-nitropyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.