missing translation for 'onlineSavingsMsg'
Learn More

(2-Hydroxy-4-methoxyphenyl)(4-methylphenyl)methanone, TRC

Product Code. 30394946
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394946 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394946 Supplier Toronto Research Chemicals Supplier No. TRCH93993850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mexenone
CAS 1641-17-4
Molecular Formula C15 H14 O3
Quantity 50 mg
Synonym Mexenone
InChI Formula InChI=1S/C15H14O3/c1-10-3-5-11(6-4-10)15(17)13-8-7-12(18-2)9-14(13)16/h3-9,16H,1-2H3
SMILES COc1ccc(C(=O)c2ccc(C)cc2)c(O)c1
IUPAC Name (2-hydroxy-4-methoxyphenyl)-(4-methylphenyl)methanone
Molecular Weight (g/mol) 242.27
Formula Weight 242.0943
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.