missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methoxybenzophenone, 98%, Thermo Scientific Chemicals

Product Code. 13438328
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-57-7
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.25
MDL Number MFCD00008387
InChI Key DXGLGDHPHMLXJC-UHFFFAOYSA-N
Synonym oxybenzone, 2-hydroxy-4-methoxybenzophenone, benzophenone-3, oxybenzon, 4-methoxy-2-hydroxybenzophenone, 2-benzoyl-5-methoxyphenol, oxibenzona, oxybenzonum, 2-hydroxy-4-methoxyphenyl phenyl methanone, anuvex
PubChem CID 4632
ChEBI CHEBI:34283
IUPAC Name (2-hydroxy-4-methoxyphenyl)-phenylmethanone
SMILES COC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >62.5°C
Color White to Yellow
Boiling Point 150°C to 160°C (5.0 mmHg)
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula HOC6H3(OCH3)COC6H5
Quantity 1 kg
Beilstein 08,312
Merck Index 15,7053
Solubility Information Solubility in water: .. Other solubilities: readily soluble in most organic solvents
Formula Weight 228.25
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-4-methoxybenzophenone, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'