missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methoxybenzophenone-5-sulfonic Acid Sodium Salt, TRC

Product Code. 30397053
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397053 2.5 mg 2.5mg
30393798 25 mg -
30402156 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397053 Supplier Toronto Research Chemicals Supplier No. TRCH5314502.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6628-37-1
Molecular Formula C14 H11 O6 S . Na
Molecular Weight (g/mol) 330.29
Synonym 2-Hydroxy-4-methoxybenzophenone-5-sulfonic Acid Sodium, Benzenesulfonic acid, 5-benzoyl-4-hydroxy-2-methoxy-, monosodium salt (9CI), Benzenesulfonic acid, 5-benzoyl-4-hydroxy-2-methoxy-, sodium salt (7CI), 2-Hydroxy-4-methoxybenzophenone-5-sodium sulfonate, 2-Hydroxy-4-methoxybenzophenone-5-sulfonic acid sodium salt, ASL 24ST, Benzophenone 5, Cyasorb UV 284, Sodium 2-hydroxy-4-methoxybenzophenone-5-sulfonate, Sodium 5-benzoyl-4-hydroxy-2-methoxybenzenesulfonate, Sodium benzophenone-2-hydroxy-4-methoxy-5-sulfonate, Sulibenzone, Uvin 1MS40, Uvistat 1121
IUPAC Name sodium;5-benzoyl-4-hydroxy-2-methoxybenzenesulfonate
SMILES [Na+].COc1cc(O)c(cc1S(=O)(=O)[O-])C(=O)c2ccccc2

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C14H12O6S.Na/c1-20-12-8-11(15)10(7-13(12)21(17,18)19)14(16)9-5-3-2-4-6-9;/h2-8,15H,1H3,(H,17,18,19);/q;+1/p-1
Formula Weight 330.0174
Chemical Name or Material 2-Hydroxy-4-methoxybenzophenone-5-sulfonic Acid Sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.