missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methoxybenzophenone-2',3',4',5',6'-d5, TRC

Product Code. 30135227
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135227 50 mg 50mg
17907309 10 mg 10mg
17928499 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30135227 Supplier Toronto Research Chemicals Supplier No. TRCO86730650MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1219798-54-5
Molecular Formula C14 2H5 H7 O3
Molecular Weight (g/mol) 233.27
Synonym Methanone, (2-hydroxy-4-methoxyphenyl)phenyl-2,3,4,5,6-d5-, Oxybenzone-d5, Oxybenzone (phenyl-D5)
IUPAC Name (2-hydroxy-4-methoxyphenyl)-(2,3,4,5,6-pentadeuteriophenyl)methanone
SMILES [2H]c1c([2H])c([2H])c(C(=O)c2ccc(OC)cc2O)c([2H])c1[2H]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H12O3/c1-17-11-7-8-12(13(15)9-11)14(16)10-5-3-2-4-6-10/h2-9,15H,1H3/i2D,3D,4D,5D,6D
Formula Weight 233.11
Percent Purity >95%
Chemical Name or Material Oxybenzone phenyl-d5
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.