missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methoxybenzaldehyde, 98%

Product Code. p-7020870
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11489833 1 g 1g
11499833 5 g 5g
11409843 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11489833

Brand: Thermo Scientific Alfa Aesar A12971.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 673-22-3 | C8H8O3 | 152.15 g/mol

It is employed in the synthesis of Schiff base ligand. It is applied as a reactant in the synthesis of LPA1R antagonists used in the inhibition of LPA-induced proliferation and contraction of normal human lung fibroblasts. Also used in the synthesis of tyrosine kinase 6 proteinase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of Schiff base ligand. It is applied as a reactant in the synthesis of LPA1R antagonists used in the inhibition of LPA-induced proliferation and contraction of normal human lung fibroblasts. Also used in the synthesis of tyrosine kinase 6 proteinase inhibitors.

Solubility
Solubility in methanol is almost transparent. Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents, bases and air/oxygen. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 673-22-3
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00003327
InChI Key WZUODJNEIXSNEU-UHFFFAOYSA-N
Synonym 4-methoxysalicylaldehyde, benzaldehyde, 2-hydroxy-4-methoxy, 4-methoxysalicyaldehyde, 2-hydroxy-4-methoxy-benzaldehyde, o-hydroxy-p-methoxybenzaldehyde, p-anisaldehyde, 2-hydroxy, salicylaldehyde, 4-methoxy, 2-hydroxy-p-anisaldehyde, 2-formyl-5-methoxyphenol, unii-2n395p88lw
PubChem CID 69600
IUPAC Name 2-hydroxy-4-methoxybenzaldehyde
SMILES COC1=CC=C(C=O)C(O)=C1

Specifications

Melting Point 40°C to 44°C
Flash Point >110°C (230°F)
Quantity 1 g
Beilstein 1072443
Sensitivity Air Sensitive
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-4-methoxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.